Läs mer
Deferasirox Methyl Ester, TRC
Produktkod. 30136627
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136627 250 mg
30138449 100 mg
30009943 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136627 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD228675250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1266741-05-2
Molekylformel C22 H17 N3 O4
Molekylvikt (g/mol) 387.39
Synonym Benzoic acid, 4-[3,5-bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]-, methyl ester, Deferasirox Methyl Ester, Methyl 4-[3,5-bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoate
IUPAC-namn methyl 4-[3,5-bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoate
LEDER COC(=O)c1ccc(cc1)n2nc(nc2c3ccccc3O)c4ccccc4O
Specifikationer
Kemiskt namn eller material Deferasirox Methyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C22H17N3O4/c1-29-22(28)14-10-12-15(13-11-14)25-21(17-7-3-5-9-19(17)27)23-20(24-25)16-6-2-4-8-18(16)26/h2-13,26-27H,1H3
Formel vikt 387.1219
Procent renhet >95%