Läs mer
Defluoro 8-Hydroxy Flumazenil, TRC
Produktkod. 30355598
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30355598 100 mg
1 options
Produktkod. 30355598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD228740100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material ethyl 8-hydroxy-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
CAS 131666-45-0
Molekylformel C15 H15 N3 O4
Kvantitet 100 mg
Synonym Ethyl 8-hydroxy-5,6-dihydro-5-methyl-6-oxo-4H-imidazol-[1,5-alpha][1,4]benzodiazepine-3-carboxylate, Flumazenil Imp. B (EP), Flumazenil Related Compound B (USP)
InChI formel InChI=1S/C15H15N3O4/c1-3-22-15(21)13-12-7-17(2)14(20)10-6-9(19)4-5-11(10)18(12)8-16-13/h4-6,8,19H,3,7H2,1-2H3
LEDER CCOC(=O)c1ncn2c1CN(C)C(=O)c3cc(O)ccc23
IUPAC-namn ethyl 8-hydroxy-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
Molekylvikt (g/mol) 301.3
Formel vikt 301.1063