Läs mer
Defluoro Dolutegravir (>85%), TRC
Produktkod. 30179595
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179595 1 mg
30179265 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2287151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1863916-88-4
Molekylformel C20 H20 F N3 O5
Molekylvikt (g/mol) 401.39
Synonym 2H-Pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide, N-[(4-fluorophenyl)methyl]-3,4,6,8,12,12a-hexahydro-7-hydroxy-4-methyl-6,8-dioxo-, (4R,12aS)-, GSK 2968453A
IUPAC-namn (4R,12aS)-N-[(4-fluorophenyl)methyl]-7-hydroxy-4-methyl-6,8-dioxo-3,4,12,12a-tetrahydro-2H-pyrido[5,6]pyrazino[2,6-b][1,3]oxazine-9-carboxamide
LEDER C[C@@H]1CCO[C@H]2CN3C=C(C(=O)NCc4ccc(F)cc4)C(=O)C(=C3C(=O)N12)O
Specifikationer
Kemiskt namn eller material 2-Desfluorodolutegravir
Kvantitet 1 mg
InChI formel InChI=1S/C20H20FN3O5/c1-11-6-7-29-15-10-23-9-14(17(25)18(26)16(23)20(28)24(11)15)19(27)22-8-12-2-4-13(21)5-3-12/h2-5,9,11,15,26H,6-8,10H2,1H3,(H,22,27)/t11-,15+/m1/s1
Formel vikt 401.1387