Läs mer
Defluoro Flunarizine Dihydrochloride, TRC
Produktkod. 30363986
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363986 50 mg
30364546 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363986 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22870050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27064-96-6
Molekylformel C26 H27 F N2 . 2 Cl H
Molekylvikt (g/mol) 459.43
Synonym Piperazine, 1-[(4-fluorophenyl)phenylmethyl]-4-(3-phenyl-2-propen-1-yl)-, hydrochloride (1:2), Piperazine, 1-[(4-fluorophenyl)phenylmethyl]-4-(3-phenyl-2-propenyl)-, dihydrochloride (9CI), Piperazine, 1-cinnamyl-4-(p-fluoro-α-phenylbenzyl)-, dihydrochloride (8CI), 1-[(RS)-(4-Fluorophenyl)phenylmethyl]-4-[(2EZ)-3-phenylprop-2-enyl]piperazine dihydrochloride, (EZ)-Defluoro-flunarizine dihydrochloride
IUPAC-namn 1-[(4-fluorophenyl)-phenylmethyl]-4-(3-phenylprop-2-enyl)piperazine;dihydrochloride
LEDER Cl.Cl.Fc1ccc(cc1)C(N2CCN(CC=Cc3ccccc3)CC2)c4ccccc4
Specifikationer
Kemiskt namn eller material Defluoro Flunarizine dihydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C26H27FN2.2ClH/c27-25-15-13-24(14-16-25)26(23-11-5-2-6-12-23)29-20-18-28(19-21-29)17-7-10-22-8-3-1-4-9-22;;/h1-16,26H,17-21H2;2*1H
Formel vikt 458.1692