Läs mer
Defluoro Hydroxy AM-694, TRC
Produktkod. 30354239
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30354239 10 mg
1 options
Produktkod. 30354239 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22873010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material AM694 N-(5-hydroxypentyl) metabolite
CAS 335160-94-6
Molekylformel C20H20INO2
Kvantitet 10 mg
Synonym Methanone, [1-(5-hydroxypentyl)-1H-indol-3-yl](2-iodophenyl)-, [1-(5-Hydroxypentyl)-1H-indol-3-yl](2-iodophenyl)methanone, AM694 N-(5-hydroxypentyl) metabolite, AM-694 N-(5-hydroxypentyl) metabolite, AM-694 N-(5-hydroxypentyl), Defluoro Hydroxy AM-694, [1-(5-Hydroxypentyl)indol-3-yl](2-iodophenyl)methanone
InChI formel InChI=1S/C20H20INO2/c21-18-10-4-2-9-16(18)20(24)17-14-22(12-6-1-7-13-23)19-11-5-3-8-15(17)19/h2-5,8-11,14,23H,1,6-7,12-13H2
LEDER OCCCCCn1cc(C(=O)c2ccccc2I)c3ccccc13
IUPAC-namn [1-(5-hydroxypentyl)indol-3-yl]-(2-iodophenyl)methanone
Molekylvikt (g/mol) 433.28
Formel vikt 433.0539