Läs mer
Defluoro-MDV 3100 (Defluoro-enzalutamide), TRC
Produktkod. 30353695
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353695 25 mg
30351258 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353695 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22859025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 915087-16-0
Molekylformel C21H17F3N4O2S
Molekylvikt (g/mol) 446.45
Synonym 4-[3-[4-Cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-4-oxo-2-thioxo-1-imidazolidinyl]-N-methylbenzamide, 4-[3-(4-Cyano-3-trifluoromethylphenyl)-5,5-dimethyl-4-oxo-2-thioxoimidazolidin-1-yl]-N-methylbenzamide
IUPAC-namn 4-[3-[4-cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-4-oxo-2-sulfanylideneimidazolidin-1-yl]-N-methylbenzamide
LEDER CNC(=O)c1ccc(cc1)N2C(=S)N(C(=O)C2(C)C)c3ccc(C#N)c(c3)C(F)(F)F
Specifikationer
Kemiskt namn eller material Defluoro-MDV 3100 (Defluoro-enzalutamide)
Kvantitet 25 mg
InChI formel InChI=1S/C21H17F3N4O2S/c1-20(2)18(30)27(15-9-6-13(11-25)16(10-15)21(22,23)24)19(31)28(20)14-7-4-12(5-8-14)17(29)26-3/h4-10H,1-3H3,(H,26,29)