Läs mer
Defluoro Paroxetine Hydrochloride, TRC
Produktkod. 30363988
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363988 100 mg
30363165 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363988 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229035100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1322626-23-2
Molekylformel C19 H22 Cl N O3
Molekylvikt (g/mol) 347.84
Synonym 3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenyl-piperidine Hydrochloride, USP Paroxetine Related Compound B
IUPAC-namn (3S)-3-(1,3-benzodioxol-5-yloxymethyl)-4-phenylpiperidine
LEDER [C@@H]1(COC2=CC=C(OCO3)C3=C2)CNCC[C@H]1C4=CC=CC=C4.Cl
Specifikationer
Kemiskt namn eller material Defluoro Paroxetine Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C19H21NO3/c1-2-4-14(5-3-1)17-8-9-20-11-15(17)12-21-16-6-7-18-19(10-16)23-13-22-18/h1-7,10,15,17,20H,8-9,11-13H2/t15-,17?/m0/s1