Learn More
ROBU VitraPOR™ Degassing Filter, Cylindrical Shape
Degassing Filter with narrow short glass tube.
Brand:  ROBU VitraPOR™ 18502
Additional Details : Weight : 0.06080kg
 View more versions of this product
Product Code. 16822976
  • 1815.60 SEK / Pack of 5
Estimated Shipment: 17-12-2021
Add to basket
Description
Description
  • For degassing with inert gases and filtration of solvents (e.g. in chromatography)
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass 3.3
12.5 mm
2
Narrow Short Glass Tube