Läs mer
Dehydro Aripiprazole, TRC
Produktkod. 17923625
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17923625 5 mg
17962715 25 mg
17954665 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17923625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2291555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 129722-25-4
Molekylformel C23 H25 Cl2 N3 O2
Molekylvikt (g/mol) 446.37
Synonym 7-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy]quinolin-2(1H)-one, Dehydroaripiprazole, Aripiprazole Related Compound G, Aripiprazole Imp E (EP), 2(1H)-Quinolinone, 7-[4-[4-(2,3-dichlorophenyl)-1-piperazinyl]butoxy]-, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-2(1H)-quinolinone, DM 14857, OPC 14857, Dehydro Aripiprazole
IUPAC-namn 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1H-quinolin-2-one
LEDER Clc1cccc(N2CCN(CCCCOc3ccc4C=CC(=O)Nc4c3)CC2)c1Cl
Specifikationer
Kemiskt namn eller material Dehydro Aripiprazole
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C23H25Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-9,16H,1-2,10-15H2,(H,26,29)
Formel vikt 445.1324
Procent renhet >95%