Läs mer
Dehydro Felodipine Ester Lactone, TRC
Produktkod. 30359558
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30359558 1 mg
1 options
Produktkod. 30359558 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2296901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dehydro Felodipine Ester Lactone
CAS 96558-27-9
Molekylformel C17 H13 Cl2 N O4
Kvantitet 1 mg
Synonym Furo[3,4-b]pyridine-3-carboxylic acid, 4-(2,3-dichlorophenyl)-5,7-dihydro-2-methyl-5-oxo-, ethyl ester, H 152/77, Ethyl 4-(2,3-Dichlorophenyl)-2-methyl-5-oxo-7H-furo[3,4-b]pyridine-3-carboxylate
InChI formel InChI=1S/C17H13Cl2NO4/c1-3-23-16(21)12-8(2)20-11-7-24-17(22)14(11)13(12)9-5-4-6-10(18)15(9)19/h4-6H,3,7H2,1-2H3
LEDER CCOC(=O)c1c(C)nc2COC(=O)c2c1c3cccc(Cl)c3Cl
IUPAC-namn ethyl 4-(2,3-dichlorophenyl)-2-methyl-5-oxo-7H-furo[3,4-b]pyridine-3-carboxylate
Molekylvikt (g/mol) 366.2
Formel vikt 365.0222