Läs mer
Dehydro Lercanidipine, TRC
Produktkod. 17913625
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17913625 10 mg
17991905 100 mg
17923295 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17913625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22922010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 887769-34-8
Molekylformel C36 H39 N3 O6
Molekylvikt (g/mol) 609.71
Synonym 2,6-dimethyl-4-(3-nitrophenyl)-3,5-Pyridinedicarboxylic acid 3-[2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl] 5-methyl ester, 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(3-nitrophenyl)-, 2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl methyl ester (9CI), Dehydro Lercanidipine, Dehydrolercanidipine
IUPAC-namn 3-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 5-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate
LEDER COC(=O)c1c(C)nc(C)c(C(=O)OC(C)(C)CN(C)CCC(c2ccccc2)c3ccccc3)c1c4cccc(c4)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Dehydro Lercanidipine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C36H39N3O6/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27/h7-19,22,30H,20-21,23H2,1-6H3
Formel vikt 609.2839
Procent renhet >95%