Läs mer
Dehydro Levetiracetam, TRC
Produktkod. 17994275
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994275 100 mg
17950465 10 mg
17960675 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD230275100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 358629-47-7
Molekylformel C8 H12 N2 O2
Molekylvikt (g/mol) 168.19
Synonym 1-Pyrrolidineacetamide, α-ethylidene-2-oxo-, (αZ)-, (αZ)-α-Ethylidene-2-oxo-1-pyrrolidineacetamide, Dehydro Levetiracetam, (2Z)-2-(2-Oxopyrrolidin-1-yl)but-2-enamide, Levetiracetam Crotonamide, Levetiracetam Imp. B (EP)
IUPAC-namn (Z)-2-(2-oxopyrrolidin-1-yl)but-2-enamide
LEDER C\C=C(/N1CCCC1=O)\C(=O)N
Specifikationer
Kemiskt namn eller material (2Z)-2-(2-Oxopyrrolidin-1-yl)but-2-enamide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C8H12N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h2H,3-5H2,1H3,(H2,9,12)/b6-2-
Formel vikt 168.0899
Procent renhet >95%