Läs mer
Dehydro Nicardipine Hydrochloride, TRC
Produktkod. 30012378
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012378 25 mg
30004286 50 mg
30004993 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012378 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22978025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1216817-27-4
Molekylformel C26 H27 N3 O6 . Cl H
Molekylvikt (g/mol) 513.97
Synonym 3-Methyl 5-[Methyl(phenylmethyl)amino]ethyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate hydrochloride, 2-[Benzyl(methyl)amino]ethyl methyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate hydrochloride, Dehydronicardipine hydrochloride
IUPAC-namn 3-O-[2-[benzyl(methyl)amino]ethyl] 5-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate;hydrochloride
LEDER Cl.COC(=O)c1c(C)nc(C)c(C(=O)OCCN(C)Cc2ccccc2)c1c3cccc(c3)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Dehydro Nicardipine Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C26H27N3O6.ClH/c1-17-22(25(30)34-4)24(20-11-8-12-21(15-20)29(32)33)23(18(2)27-17)26(31)35-14-13-28(3)16-19-9-6-5-7-10-19;/h5-12,15H,13-14,16H2,1-4H3;1H
Formel vikt 513.1667