Läs mer
Dehydro Silodosin, TRC
Produktkod. 30377072
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30377072 10 mg
30373447 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30377072 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44645010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 175870-21-0
Molekylformel C25H30F3N3O4
Molekylvikt (g/mol) 493.52
Synonym 1-(3-Hydroxypropyl)-5-[(2R)-2-[[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl]amino]propyl]-1H-indole-7-carboxamide
IUPAC-namn 1-(3-hydroxypropyl)-5-[(2R)-2-[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethylamino]propyl]indole-7-carboxamide
LEDER C[C@H](Cc1cc(C(=O)N)c2c(ccn2CCCO)c1)NCCOc3ccccc3OCC(F)(F)F
Specifikationer
Kemiskt namn eller material Silodosin Dehydro
Kvantitet 10 mg
InChI formel InChI=1S/C25H30F3N3O4/c1-17(30-8-12-34-21-5-2-3-6-22(21)35-16-25(26,27)28)13-18-14-19-7-10-31(9-4-11-32)23(19)20(15-18)24(29)33/h2-3,5-7,10,14-15,17,30,32H,4,8-9,11-13,16H2,1H3,(H2,29,33)/t17-/m1/s1
Formel vikt 493.2188