missing translation for 'onlineSavingsMsg'
Läs mer

Dehydro Warfarin, TRC

Produktkod. 30367881
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367881 25 mg
30365656 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30367881 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23058525MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 67588-18-5
Molekylformel C19H14O4
Molekylvikt (g/mol) 306.31
Synonym Dehydrowarfarin, 4-Hydroxy-3-(3-oxo-1-phenyl-1-buten-1-yl)-2H-1-benzopyran-2-one, NSC 289346
LEDER CC(=O)\C=C(\C1=C(O)c2ccccc2OC1=O)/c3ccccc3

Specifications

Kemiskt namn eller material Dehydro Warfarin
Kvantitet 25 mg
InChI formel InChI=1S/C19H14O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-11,21H,1H3/b15-11+
Formel vikt 306.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.