Läs mer
Dehydroabietal, TRC
Produktkod. 30135924
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30135924 5 mg -
30012907 10 mg -
30010922 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135924 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2290985MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13601-88-2
Molekylformel C20 H28 O
Molekylvikt (g/mol) 284.44
Synonym (1R,4aS,10aR)-1,2,3,4,4a,9,10,10a-Octahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxaldehyde, 13-Isopropylpodocarpa-8,11,13-triene-15-aldehyde, Dehydroabietic Aldehyde, Dehydroabietinal
IUPAC-namn (1R,4aS,10aR)-7-isopropyl-1,4a-dimethyl-2,3,4,9,10,10a-hexahydrophenanthrene-1-carbaldehyde
LEDER CC(C)c1ccc2c(CC[C@H]3[C@@](C)(CCC[C@]23C)C=O)c1
Specifikationer
Kemiskt namn eller material Dehydroabietal
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C20H28O/c1-14(2)15-6-8-17-16(12-15)7-9-18-19(3,13-21)10-5-11-20(17,18)4/h6,8,12-14,18H,5,7,9-11H2,1-4H3/t18-,19-,20+/m0/s1
Formel vikt 284.214
Procent renhet >95%