Läs mer
Dehydroabietic Acid, TRC
Produktkod. 17930585
Change view
Kvantitet:
1 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17930585 50 mg
30133919 100 mg
17941845 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17930585 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22908550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1740-19-8
Molekylformel C20 H28 O2
Molekylvikt (g/mol) 300.44
Synonym 1-Phenanthrenecarboxylic acid, 1,2,3,4,4a,9,10,10a-octahydro-1,4a-dimethyl-7-(1-methylethyl)-, (1R,4aS,10aR)-, 1-Phenanthrenecarboxylic acid, 1,2,3,4,4a,9,10,10a-octahydro-1,4a-dimethyl-7-(1-methylethyl)-, [1R-(1α,4aβ,10aα)]-, Abietic acid, dehydro- (6CI), Podocarpa-8,11,13-trien-15-oic acid, 13-isopropyl- (7CI,8CI), (1R,4aS,10aR)-1,2,3,4,4a,9,10,10a-Octahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxylic acid, (+)-Dehydroabietic acid, 7-Callitrisic acid, Abieta-8,11,13-trien-18-oic acid, Dehydroabietic acid, NSC 2952, Resigral 52
IUPAC-namn (1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-1-carboxylic acid
LEDER CC(C)c1ccc2c(CC[C@H]3[C@@](C)(CCC[C@]23C)C(=O)O)c1
Specifikationer
Kemiskt namn eller material Dehydroabietic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C20H28O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h6,8,12-13,17H,5,7,9-11H2,1-4H3,(H,21,22)/t17-,19-,20-/m1/s1
Formel vikt 300.2089
Procent renhet >95%