Learn More
Dehydroabietinol, TRC
Product Code. 30359992
Change view
Click to view available options
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30359992 1 g
30349960 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30359992 Supplier Toronto Research Chemicals Supplier No. TRCD2290951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3772-55-2
Molekylformel C20 H34 O
Molekylvikt (g/mol) 290.48
Synonym (1R,4aS,10aR)-1,2,3,4,4a,9,10,10a-Octahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenemethanol, 13-Isopropylpodocarpa-8,11,13-trien-15-ol, Dehydroabeityl Alcohol, Dehydroabietol, Dehydroabietyl alcohol, Pomiferin A, ar-Abietatrienol, [1R-(1α,4aβ,10aα)]-1,2,3,4,4a,9,10,10a-Octahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenemethanol
IUPAC-namn (1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,7,9,10,10a-decahydrophenanthren-1-yl)methanol
LEDER CC(C)C1CCC2C(=C1)CCC3C(C)(CO)CCCC23C
Specifications
Kemiskt namn eller material Dehydroabietinol
Kvantitet 1 g
InChI formel InChI=1S/C20H34O/c1-14(2)15-6-8-17-16(12-15)7-9-18-19(3,13-21)10-5-11-20(17,18)4/h12,14-15,17-18,21H,5-11,13H2,1-4H3
Formel vikt 290.261