missing translation for 'onlineSavingsMsg'
Läs mer

Dehydroacetic Acid, Sigma-Aldrich™

Product Code. 15851611
Change view
Click to view available options
Kvantitet:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
15851611 250 g 250g
15841611 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15851611 Supplier Sigma Aldrich Supplier No. 8220110250
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-45-6

Specifications

Smältpunkt 109°C to 111°C
Kokpunkt 270°C/1013 hPa
MDL-nummer MFCD00066709
Kvantitet 250 g
Synonym Dehydroacetic acid, 3-Acetyl-3,4-dihydro-6-methyl-2H-pyran-2,4-dione, Dehydroacetic acid
Molekylvikt (g/mol) 168.15
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.