Läs mer
Dehydrocholic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dehydrocholic Acid |
| CAS | 81-23-2 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C24 H34 O5 |
| Kvantitet | 5 g |
| Synonym | Dehydrocholic Acid, Cholan-24-oic acid, 3,7,12-trioxo-, (5β)-, 5β-Cholan-24-oic acid, 3,7,12-trioxo- (8CI), 3,7,12-Tri-keto-5β-Cholan-24-oic acid, 3,7,12-Triketo-5β-cholanic acid, 3,7,12-Triketo-5β-cholanoic acid, 3,7,12-Triketocholanic acid, 3,7,12-Trioxo-5β-cholan-24-oic acid, 3,7,12-Trioxo-5β-cholanic acid, 3,7,12-Trioxocholanic acid, Acolen, Bilidren, Bilostat, Cholagon, Cholan DH, Cholepatin, Cholic acid, dehydro-, Cholimed, Chologon, DHC, Decholin, Dehychol, Dehycon, Dehydrocholic acid, Dehystolin, Deidrocolico Vita, Didocol, Didrocolo, Dilabil, Drenobyl, Erebile, Felacrinos, Hydrochol, Hykolex, Ketocholanic acid, NSC 8796, Novocolin, Oxycholin, Procholon, Sanocholen, Triketocholanic acid |
| InChI formel | InChI=1S/C24H34O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-14,16-18,22H,4-12H2,1-3H3,(H,28,29)/t13-,14+,16-,17+,18+,22+,23+,24-/m1/s1 |
| LEDER | C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@H]3[C@H](CC(=O)[C@]12C)[C@@]4(C)CCC(=O)C[C@H]4CC3=O |
| IUPAC-namn | (4R)-4-[(5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3,7,12-trioxo-1,2,4,5,6,8,9,11,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid |
| Molekylvikt (g/mol) | 402.52 |
| Visa mer |