Läs mer
Dehydronitroso Nifedipine, TRC
Produktkod. 30355039
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355039 100 mg
30365382 25 mg
30362660 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355039 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229890100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50428-14-3
Molekylformel C17 H16 N2 O5
Molekylvikt (g/mol) 328.32
Synonym Dimethyl 2,6-Dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate, Nifedipine Imp. B (EP), Nitrosophenylpyridine Analogue of Nifedipine
IUPAC-namn dimethyl 2,6-dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate
LEDER COC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2ccccc2N=O
Specifikationer
Kemiskt namn eller material Dimethyl-2,6-Dimethyl-4-(2-Nitrosophenyl) Pyridine-3,5-Dicarboxylate
Kvantitet 100 mg
InChI formel InChI=1S/C17H16N2O5/c1-9-13(16(20)23-3)15(11-7-5-6-8-12(11)19-22)14(10(2)18-9)17(21)24-4/h5-8H,1-4H3
Formel vikt 328.1059