Läs mer
Dehydronitrosonisoldipine, TRC
Produktkod. 30362856
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362856 50 mg
30365851 100 mg
30361703 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23031050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 87375-91-5
Molekylformel C20H22N2O5
Molekylvikt (g/mol) 370.4
Synonym 2,6-Dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic Acid 3-Methyl 5-(2-Methylpropyl) Ester, 2,6-Dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic Acid Methyl 2-Methylpropyl Ester, 5-Methyl 3-(2-Methylprop-3-yl) 2,6-Dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate, Nisoldipine Impurity D
LEDER COC(=O)c1c(C)nc(C)c(C(=O)OCC(C)C)c1c2ccccc2N=O
Specifikationer
Kemiskt namn eller material Dehydronitrosonisoldipine
Kvantitet 50 mg
InChI formel InChI=1S/C20H22N2O5/c1-10(2)13(5)27-20(25)17-12(4)21-11(3)16(19(23)24)18(17)14-8-6-7-9-15(14)22-26/h6-10,13H,1-5H3,(H,23,24)/p-1
Formel vikt 370.1529