Läs mer
Dehydropinaverium Bromide, TRC
Produktkod. 30004917
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004917 250 mg
30006308 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004917 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB683795250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1235355-01-7
Molekylformel C26 H39 Br N O4 . Br
Molekylvikt (g/mol) 589.4
Synonym (1R,5S)-4-[(2-Bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl)ethoxy]ethyl]morpholinium Bromide (1:1), Morpholinium, 4-[(2-bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl)ethoxy]ethyl]-, bromide (1:1)
IUPAC-namn 4-[(2-bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-[(1R,5S)-6,6-dimethyl-4-bicyclo[3.1.1]hept-3-enyl]ethoxy]ethyl]morpholin-4-ium;bromide
LEDER [Br-].COc1cc(Br)c(C[N+]2(CCOCCC3=CC[C@@H]4C[C@H]3C4(C)C)CCOCC2)cc1OC
Specifikationer
Kemiskt namn eller material (1R,5S)-4-[(2-Bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl)ethoxy]ethyl]morpholinium Bromide
Kvantitet 250 mg
InChI formel InChI=1S/C26H39BrNO4.BrH/c1-26(2)21-6-5-19(22(26)16-21)7-11-31-12-8-28(9-13-32-14-10-28)18-20-15-24(29-3)25(30-4)17-23(20)27;/h5,15,17,21-22H,6-14,16,18H2,1-4H3;1H/q+1;/p-1/t21-,22-;/m1./s1
Formel vikt 587.1246