Läs mer
Dehydroxy Mirabegron Hydrochloride Salt, TRC
Produktkod. 30135177
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135177 500 mg
30133014 250 mg
30004848 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135177 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD230170500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1581284-79-8
Molekylformel C21H24N4OS . (HCl)
Molekylvikt (g/mol) 417
Synonym 2-Amino-N-[4-[2-[(2-phenylethyl)amino]ethyl]phenyl]-4-Thiazoleacetamide Hydrochloride Salt;
LEDER Cl.Nc1nc(CC(=O)Nc2ccc(CCNCCc3ccccc3)cc2)cs1
Specifikationer
Kemiskt namn eller material Dehydroxy Mirabegron Hydrochloride Salt
Kvantitet 500 mg
InChI formel InChI=1S/C21H24N4OS.ClH/c22-21-25-19(15-27-21)14-20(26)24-18-8-6-17(7-9-18)11-13-23-12-10-16-4-2-1-3-5-16;/h1-9,15,23H,10-14H2,(H2,22,25)(H,24,26);1H
Formel vikt 416.1438