Läs mer
Dehydroxy Ractopamine-d6 (Major) Hydrochloride Salt, TRC
Produktkod. 30351777
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30351777 1 mg
1 options
Produktkod. 30351777 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2306021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dehydroxy Ractopamine-d6 Hydrochloride
CAS 1346599-88-9
Molekylformel C18H18D6ClNO2
Kvantitet 1 mg
Synonym 4-[3-[[2-(4-Hydroxyphenyl)ethyl]amino]butyl]phenol-d6 Hydrochloride Salt
InChI formel InChI=1S/C18H23NO2.ClH/c1-14(2-3-15-4-8-17(20)9-5-15)19-13-12-16-6-10-18(21)11-7-16;/h4-11,14,19-21H,2-3,12-13H2,1H3;1H/i1D3,2D2,14D;
LEDER OC1=CC=C(CCNC([2H])(C([2H])([2H])[2H])C([2H])([2H])CC2=CC=C(O)C=C2)C=C1.Cl
Molekylvikt (g/mol) 327.88