Läs mer
Dehydroxy Ractopamine, TRC
Produktkod. 30351776
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351776 50 mg
30353345 25 mg
30368150 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30351776 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23060050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246816-72-7
Molekylformel C18H23NO2
Molekylvikt (g/mol) 285.38
Synonym Dehydroxy Optaflexx, Dehydroxy Paylean, Dehydroxy Topmax 9, 4-[3-[[2-(4-Hydroxyphenyl)ethyl]amino]butyl]phenol
LEDER CC(CCc1ccc(O)cc1)NCCc2ccc(O)cc2
Specifikationer
Kemiskt namn eller material Dehydroxy Ractopamine
Kvantitet 50 mg
InChI formel InChI=1S/C18H23NO2/c1-14(2-3-15-4-8-17(20)9-5-15)19-13-12-16-6-10-18(21)11-7-16/h4-11,14,19-21H,2-3,12-13H2,1H3
Formel vikt 285.1729