missing translation for 'onlineSavingsMsg'
Läs mer

Dehydroxyamino Oxamflatin Acid, TRC

Product Code. 30367365
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30367365 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367365 Supplier Toronto Research Chemicals Supplier No. TRCD22991010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Dehydroxyamino Oxamflatin Acid
CAS 151720-90-0
Molekylformel C17H13NO4S
Kvantitet 10 mg
Synonym (2E)-5-[3-[(Phenylsulfonyl)amino]phenyl]-2-penten-4-ynoic Acid
InChI formel InChI=1S/C17H13NO4S/c19-17(20)12-5-4-7-14-8-6-9-15(13-14)18-23(21,22)16-10-2-1-3-11-16/h1-3,5-6,8-13,18H,(H,19,20)/b12-5+
LEDER OC(=O)\C=C\C#Cc1cccc(NS(=O)(=O)c2ccccc2)c1
IUPAC-namn (E)-5-[3-(benzenesulfonamido)phenyl]pent-2-en-4-ynoic acid
Molekylvikt (g/mol) 327.35
Formel vikt 327.0565
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.