missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16448-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein with one SAP domain. This protein binds to cruciform and super helical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases. Two transcript variants encoding different isoforms have been found for this gene.Specifications
DEK | |
Polyclonal | |
Unconjugated | |
DEK | |
D6S231E, DEK, DEK oncogene, Protein DEK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110052, 306817, 7913 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35659, Q6AXS3, Q7TNV0 | |
DEK | |
DEK Fusion Protein Ag8225 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |