Läs mer
Delavirdine, TRC
Produktkod. 30358547
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358547 25 mg
30353055 10 mg
30356317 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358547 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23062525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136817-59-9
Molekylformel C22 H28 N6 O3 S
Molekylvikt (g/mol) 456.56
Synonym Delavirdine, N-[2-[[4-[3-[(1-Methylethyl)amino]-2-pyridinyl]-1-piperazinyl]carbonyl]-1H-indol-5-yl]methanesulfonamide (ACI), Piperazine, 1-[3-[(1-methylethyl)amino]-2-pyridinyl]-4-[[5-[(methylsulfonyl)amino]-1H-indol-2-yl]carbonyl]- (9CI), 1-[3-(Isopropylamino)-2-pyridyl]-4-[(5-methanesulfonamidoindol-2-yl)carbonyl]piperazine, 2-[4-(5-Methanesulfonamido-1H-indol-2-ylcarbonyl)-1-piperazinyl]-N-(1-methylethyl)-3-pyridinamine, BHAP-U 90152, Delavirdine, U 90152
IUPAC-namn N-[2-[4-[3-(propan-2-ylamino)pyridin-2-yl]piperazine-1-carbonyl]-1H-indol-5-yl]methanesulfonamide
LEDER CC(C)Nc1cccnc1N2CCN(CC2)C(=O)c3cc4cc(NS(=O)(=O)C)ccc4[nH]3
Specifikationer
Kemiskt namn eller material Delavirdine
Kvantitet 25 mg
InChI formel InChI=1S/C22H28N6O3S/c1-15(2)24-19-5-4-8-23-21(19)27-9-11-28(12-10-27)22(29)20-14-16-13-17(26-32(3,30)31)6-7-18(16)25-20/h4-8,13-15,24-26H,9-12H2,1-3H3
Formel vikt 456.1944