Läs mer
Delmadinone, TRC
Produktkod. 30361667
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361667 10 mg
30366748 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361667 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23065510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Delmadinone
CAS 15262-77-8
Molekylformel C21 H25 Cl O3
Kvantitet 10 mg
Synonym Pregna-1,4,6-triene-3,20-dione, 6-chloro-17-hydroxy-, 6-Chloro-17-hydroxypregna-1,4,6-triene-3,20-dione, 6-Chloro-17α-hydroxypregna-1,4,6-triene-3,20-dione, Delmadinone, δ1-Chlormadinone
InChI formel InChI=1S/C21H25ClO3/c1-12(23)21(25)9-6-16-14-11-18(22)17-10-13(24)4-7-19(17,2)15(14)5-8-20(16,21)3/h4,7,10-11,14-16,25H,5-6,8-9H2,1-3H3/t14?,15-,16-,19+,20-,21-/m0/s1
LEDER CC(=O)[C@@]1(O)CC[C@H]2C3C=C(Cl)C4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn (9S,10R,13S,14S,17R)-17-acetyl-6-chloro-17-hydroxy-10,13-dimethyl-9,11,12,14,15,16-hexahydro-8H-cyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 360.87
Formel vikt 360.1492