Läs mer
Delphinidin 3,5-Diglucoside (90%), TRC
Produktkod. 30357762
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30357762 5 mg
1 options
Produktkod. 30357762 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2146655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Delphinidin-3,5-Diglucoside Chloride (Delphin Chloride)
CAS 17670-06-3
Molekylformel C27 H31 O17 . Cl
Kvantitet 5 mg
Synonym 1-Benzopyrylium, 3,5-bis(β-D-glucopyranosyloxy)-7-hydroxy-2-(3,4,5-trihydroxyphenyl)-, chloride (1:1), 1-Benzopyrylium, 3,5-bis(β-D-glucopyranosyloxy)-7-hydroxy-2-(3,4,5-trihydroxyphenyl)-, chloride (9CI), Delphin (6CI,7CI,8CI), Awobana, Awobanin A, C.I. 75190, C.I. Natural Blue 3, Delphinidin 3,5-di-O-β-D-glucopyranoside, Delphinidin 3,5-diglucoside, Delphinidin diglucoside, Diglucosyl-3,5-delphinidin
InChI formel InChI=1S/C27H30O17.ClH/c28-6-16-19(34)21(36)23(38)26(43-16)41-14-4-9(30)3-13-10(14)5-15(25(40-13)8-1-11(31)18(33)12(32)2-8)42-27-24(39)22(37)20(35)17(7-29)44-27;/h1-5,16-17,19-24,26-29,34-39H,6-7H2,(H3-,30,31,32,33);1H/t16-,17-,19-,20-,21+,22+,23-,24-,26-,27-;/m1./s1
LEDER [Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc(O)cc3[o+]c2c5cc(O)c(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[7-hydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-(3,4,5-trihydroxyphenyl)chromenylium-5-yl]oxyoxane-3,4,5-triol;chloride
Molekylvikt (g/mol) 662.98
Formel vikt 662.125