Läs mer
Delphinidin Chloride, TRC
Produktkod. 30356075
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30356075 2.5 mg
1 options
Produktkod. 30356075 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2306802.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Delphinidin Chloride
CAS 528-53-0
Molekylformel C15 H11 O7 . Cl
Kvantitet 2.5 mg
Synonym 1-Benzopyrylium, 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-, chloride (1:1), 1-Benzopyrylium, 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-, chloride (9CI), Flavylium, 3,3',4',5,5',7-hexahydroxy-, chloride (8CI), 3,3',4',5,5',7-Hexahydroxy-2-phenylbenzopyrylium chloride, 3,3',4',5,5',7-Hexahydroxyflavylium chloride, Delfinidol chloride, Delphinidin, Delphinidin chloride, Delphinidine, Delphinidol, Ephdine, IdB 1056
InChI formel InChI=1S/C15H10O7.ClH/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6;/h1-5H,(H5-,16,17,18,19,20,21);1H
LEDER [Cl-].Oc1cc(O)c2cc(O)c([o+]c2c1)c3cc(O)c(O)c(O)c3
IUPAC-namn 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
Molekylvikt (g/mol) 338.7
Formel vikt 338.0193