Läs mer
delta-1,2,3,4,5,6-Hexachlorocyclohexane (Technical Grade), TRC
Produktkod. 30398985
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30398985 50 mg
1 options
Produktkod. 30398985 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29080350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material delta-HCH
CAS 319-86-8
Molekylformel C6 H6 Cl6
Kvantitet 50 mg
Synonym (1α,2α,3α,4β,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane, Delta-BHC, Δ-1,2,3,4,5,6-Hexachlorocyclohexane, Δ-666, Δ-BHC, Δ-Benzene hexachloride, Δ-HCH, Δ-Hexachlorocyclohexane, Δ-Lindane
InChI formel InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5-,6-
LEDER Cl[C@@H]1[C@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@H]1Cl
IUPAC-namn 1,2,3,4,5,6-hexachlorocyclohexane
Molekylvikt (g/mol) 290.83
Formel vikt 287.8601