Läs mer
delta-5(6)-Norethindrone Acetate, TRC
Produktkod. 30428516
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30428516 50 mg
1 options
Produktkod. 30428516 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN67601550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Oxo-19-nor-17alpha-pregn-5-en-20-yn-17-yl Acetate
CAS 1175129-26-6
Molekylformel C22 H28 O3
Kvantitet 50 mg
Synonym 3-Oxo-19-nor-17alpha-pregn-5-en-20-yn-17-yl Acetate, Norethisterone Acetate Imp. C (EP)
InChI formel InChI=1S/C22H28O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,5,17-20H,6-13H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1
LEDER CC(=O)O[C@]1(CC[C@H]2[C@@H]3CC=C4CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
IUPAC-namn [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,4,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molekylvikt (g/mol) 340.46
Formel vikt 340.2038