Läs mer
delta2-Cefuroxime, TRC
Produktkod. 30367595
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367595 50 mg
30358661 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC24806050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 229499-08-5
Molekylformel C16 H16 N4 O8 S
Molekylvikt (g/mol) 424.38
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-[[(2Z)-2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, (6R,7R)- (9CI), (6R,7R)-3-[[(Aminocarbonyl)oxy]methyl]-7-[[(2Z)-2-(2-furanyl)-2-(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid
IUPAC-namn (6R,7R)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(2-furyl)-2-methoxyimino-acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid
LEDER CO\N=C(/C(=O)N[C@H]1[C@H]2SC=C(COC(=O)N)C(N2C1=O)C(=O)O)\c3occc3
Specifikationer
Kemiskt namn eller material delta2-Cefuroxime
Kvantitet 50 mg
InChI formel InChI=1S/C16H16N4O8S/c1-26-19-9(8-3-2-4-27-8)12(21)18-10-13(22)20-11(15(23)24)7(5-28-16(17)25)6-29-14(10)20/h2-4,6,10-11,14H,5H2,1H3,(H2,17,25)(H,18,21)(H,23,24)/b19-9-/t10-,11?,14-/m1/s1
Formel vikt 424.069