Läs mer
delta7,9(11)-Dexamethasone, TRC
Produktkod. 17992965
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17992965 10 mg
17920545 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17992965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29878510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1809224-82-5
Molekylformel C22 H26 O4
Molekylvikt (g/mol) 354.44
Synonym 17,21-Dihydroxy-16α-methylpregna-1,4,7,9(11)-tetraene-3,20-dione, Dexamethason Impurity K
IUPAC-namn (10S,13S,14S,16R,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-12,14,15,16-tetrahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER C[C@@H]1C[C@H]2C3=CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 17,21-Dihydroxy-16a-methylpregna-1,4,7,9(11)-tetraene-3,20-dione
Kvantitet 10 mg
InChI formel InChI=1S/C22H26O4/c1-13-10-18-16-5-4-14-11-15(24)6-8-20(14,2)17(16)7-9-21(18,3)22(13,26)19(25)12-23/h5-8,11,13,18,23,26H,4,9-10,12H2,1-3H3/t13-,18+,20+,21+,22+/m1/s1
Formel vikt 354.1831