Läs mer
delta7-Avenasterol (E/Z mixture), TRC
Produktkod. 17968814
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17968814 0.5 mg 0.5mg
17919024 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17968814 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7948250.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23290-26-8
Molekylformel C29 H48 O
Synonym (3beta,5alpha,24Z)-Stigmasta-7,24(28)-dien-3-ol∣(24Z)-24-Ethyl-5alpha-cholesta-7,24(28)-dien-3beta-ol∣(24Z)-5alpha-Stigmasta-7,24(28)-dien-3beta-ol∣(Z)-5alpha-Stigmasta-7,24(28)-dien-3beta-ol∣24Z-Ethylidenelathosterol∣3beta-Hydroxy-5alpha-stigmasta-7,24(28)Z-diene∣5alpha-Stigmasta-7,24(28)-dien-3-ol∣5alpha-Stigmasta-7,Z-24(28)-diene-3beta-ol∣Avenasterol∣Z-24-Ethylidene-5alpha-cholest-7-en-3beta-ol
Specifikationer
Kemiskt namn eller material delta7-Avenasterol (E/Z mixture)
Kvantitet 0.5 mg
Frakt skick Room Temperature
Formel vikt 412.3705
MDL-nummer MFCD29918449
Renhet >95% (HPLC)
Molekylvikt (g/mol) 412.69, 412.70
InChI-nyckel MCWVPSBQQXUCTB-OQTIOYDCSA-N
LEDER [H][C@@]1(CC[C@@]2([H])C3=CC[C@@]4([H])C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C)[C@H](C)CC\C(=C\C)C(C)C
IUPAC-namn (1R,3aR,5aS,7S,9aS,9bR,11aR)-9a,11a-dimethyl-1-[(2R,5Z)-5-(propan-2-yl)hept-5-en-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol