Läs mer
Deltamethrin, TRC
Produktkod. 30008708
Change view
Kvantitet:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008708 1 g
30004436 10 g
18060250 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008708 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2307001G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52918-63-5
Molekylformel C22 H19 Br2 N O3
Molekylvikt (g/mol) 505.2
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1R-[1α(S*),3α]]-, (S)-Cyano(3-phenoxyphenyl)methyl (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropanecarboxylate, Alpha Max, AquaK'othrine, Battalion, Butox, Cislin, Crackdown, Decamethrin, Decamethrine, Decis, Decis 0.5ULV, Decis 1.5ULV, Decis 2.5EC, Decis 2.5ULV, Decis Expert, Decis Extra, Decis Flow, Decis Jet, Decis Super, Decis flow 2.5, Decistab, Decistab TB, Delta Gold, Delta Gold (insecticide), DeltaGard, Deltacid, Deltadust, Deltagran, Deltamethrin, Ezeetab, FMC 45498, Glossinex 200, IPO 8831, K-O Tab, K-Obiol, K-Obiol 2P, K-Othrin, K-Othrine, K-Othrine Flow 25, K-Othrine Polyzone, NRDC 161, New Musigie, Pendelta, Phagase 1, RU 22974, RUP 987, Scalibor, Spritex antiwasp powder, Suspend, Timbertreat DM 5, Ultratec, Vectocid, Zorcis
IUPAC-namn [(S)-cyano-(3-phenoxyphenyl)methyl] (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropane-1-carboxylate
LEDER CC1(C)[C@@H](C=C(Br)Br)[C@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2
Specifikationer
Kemiskt namn eller material Deltamethrin
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18+,20-/m0/s1
Formel vikt 502.9732
Procent renhet >95%