Läs mer
Deltarasin hydrochloride, MedChemExpress
Produktkod. 16499337
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16499337 5 mg -
16442867 50 mg -
16416798 10 mg 10mg
16406916 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16499337 Leverantör Medchem Express Leverantörsnummer HY15747A
MedChemExpress Deltarasin hydrochloride is an inhibitor of KRAS-PDEδinteraction with Kd of 38 nM for binding to purified PDEδ.
Deltarasin hydrochloride is an inhibitor of KRAS-PDEδinteraction with Kd of 38 nM for binding to purified PDEδ.
Specifikationer
Kemiskt namn eller material Deltarasin hydrochloride
CAS 1613404-76-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C40H38ClN5O
Kvantitet 5 mg
Löslighetsinformation DMSO : ≥ 52 mg/mL (81.22 mM) ∣H2O : 50 mg/mL (78.10 mM; Need ultrasonic)
LEDER [H]Cl.C1(C2=CC=C(OC[C@@H](N3C4=CC=CC=C4N=C3C5=CC=CC=C5)C6CCNCC6)C=C2)=NC7=CC=CC=C7N1CC8=CC=CC=C8
Molekylvikt (g/mol) 640.22
Formel vikt 640.22
Procent renhet 99.97%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease