Läs mer
Demecarium Bromide, TRC
Produktkod. 30360699
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360699 50 mg
30363890 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23072050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56-94-0
Molekylformel C32 H52 N4 O4 . 2 Br
Molekylvikt (g/mol) 716.59
Synonym Benzenaminium, 3,3'-[1,10-decanediylbis[(methylimino)carbonyloxy]]bis[N,N,N-trimethyl-, bromide (1:2), (m-Hydroxyphenyl)trimethylammonium bromide decamethylenebis[methylcarbamate] (6CI,7CI), Ammonium, (m-hydroxyphenyl)trimethyl-, bromide, decamethylenebis[methylcarbamate] (2:1) (8CI), Benzenaminium, 3,3'-[1,10-decanediylbis[(methylimino)carbonyloxy]]bis[N,N,N-trimethyl-, dibromide (9CI), BC 48, Decamethylenebis(m-dimethylaminophenyl-N-methylcarbamate) dimethobromide, Decamethylenebis[N-methylcarbamic acid m-dimethylaminophenyl ester] bromomethylate, Demecarium bromide, Demekarium bromide, Demekastigmine bromide, Frumtosnil, Humorsol, N,N'-Bis[3-trimethylammoniumphenoxycarbonyl]-N,N'-dimethyldecamethylenediamine dibromide, Tonilen, Tosmicil, Tosmilen, Tosmilene, Visumatic, Visumiotic
IUPAC-namn trimethyl-[3-[methyl-[10-[methyl-[3-(trimethylazaniumyl)phenoxy]carbonylamino]decyl]carbamoyl]oxyphenyl]azanium;dibromide
LEDER [Br-].[Br-].CN(CCCCCCCCCCN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C)C(=O)Oc2cccc(c2)[N+](C)(C)C
Specifikationer
Kemiskt namn eller material Demecarium Bromide
Kvantitet 50 mg
InChI formel InChI=1S/C32H52N4O4.2BrH/c1-33(31(37)39-29-21-17-19-27(25-29)35(3,4)5)23-15-13-11-9-10-12-14-16-24-34(2)32(38)40-30-22-18-20-28(26-30)36(6,7)8;;/h17-22,25-26H,9-16,23-24H2,1-8H3;2*1H/q+2;;/p-2
Formel vikt 714.2355