missing translation for 'onlineSavingsMsg'
Learn More

Demecolcine, ≥98%, MP Biomedicals™
BIOPHARMA

Catalog Number p-4405482
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
This item is not returnable. View return policy
477-30-5
C21H25NO5
371.43
MFCD00075459
NNJPGOLRFBJNIW-UHFFFAOYNA-N
demecolcine, colcemid, colchamine, --demecolcine, reichstein's f, colcemide, demecolcin, desmecolcine, santavy's substance f, --colchamine
220401
CHEBI:4393
CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12
This item is not returnable. View return policy
477-30-5
C21H25NO5
371.43
MFCD00075459
NNJPGOLRFBJNIW-UHFFFAOYNA-N
demecolcine, colcemid, colchamine, --demecolcine, reichstein's f, colcemide, demecolcin, desmecolcine, santavy's substance f, --colchamine
220401
CHEBI:4393
CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12
Color Yellow
Assay Percent Range >98%
Quantity 1 mg
IUPAC Name 3,4,5,14-tetramethoxy-10-(methylamino)tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-13-one
Formula Weight 371.4
Physical Form Powder
Chemical Name or Material Demecolcine, Antineoplastic agent
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'