Läs mer
Demephion, TRC
Produktkod. 30365605
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30365605 100 mg
1 options
Produktkod. 30365605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD231735100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Demephion
CAS 8065-62-1
Molekylformel 2 C5 H13 O3 P S2
Kvantitet 100 mg
Synonym Demephion, Phosphorothioic acid, O,O-dimethyl O-[2-(methylthio)ethyl] ester, mixt. with O,O-dimethyl S-[2-(methylthio)ethyl] phosphorothioate (9CI, ACI) Phosphorothioic acid, O,O-dimethyl S-[2-(methylthio)ethyl] ester, mixt. contg. (9CI), Atlasetox, Demephion, Methy
InChI formel InChI=1S/2C5H13O3PS2/c1-7-9(6,8-2)11-5-4-10-3;1-6-9(10,7-2)8-4-5-11-3/h2*4-5H2,1-3H3
LEDER COP(=S)(OC)OCCSC.COP(=O)(OC)SCCSC
IUPAC-namn dimethoxy-(2-methylsulfanylethoxy)-sulfanylidene-λ^{5}-phosphane;1-dimethoxyphosphorylsulfanyl-2-methylsulfanylethane
Molekylvikt (g/mol) 432.517
Formel vikt 432.009