Läs mer
Demethyl Curcumin, TRC
Produktkod. 30359788
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30359788 50 mg
1 options
Produktkod. 30359788 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23086550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Demethyl Curcumin
CAS 149732-51-4
Molekylformel C20H18O6
Kvantitet 50 mg
Synonym (1E,6E)-1-(3,4-Dihydroxyphenyl)-7-(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione
InChI formel InChI=1S/C20H18O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)12-15(21)6-2-13-4-8-17(23)19(25)10-13/h2-11,23-25H,12H2,1H3/b6-2+,7-3+
LEDER COC1=C(C=CC(=C1)/C=C/C(=O)CC(=O)/C=C/C2=CC(=C(C=C2)O)O)O
Molekylvikt (g/mol) 354.35
Formel vikt 354.1103