missing translation for 'onlineSavingsMsg'
Learn More

Demethylcyproheptadine Hydrochloride, Mikromol™

Product Code. 30050438
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30050438 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050438 Supplier Mikromol Supplier No. MM0609.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Demethylcyproheptadine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Demethylcyproheptadine Hydrochloride
CAS 4331-45-7
Analyt- eller komponentnamn Demethylcyproheptadine Hydrochloride
Frakt skick Room Temperature
Molekylformel C20 H19 N . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Demethylcyproheptadine Hydrochloride
InChI formel InChI=1S/C20H19N.ClH/c1-3-7-18-15(5-1)9-10-16-6-2-4-8-19(16)20(18)17-11-13-21-14-12-17;/h1-10,21H,11-14H2;1H
LEDER Cl.C1CC(=C2c3ccccc3C=Cc4ccccc24)CCN1
IUPAC-namn 4-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)piperidine;hydrochloride
Molekylvikt (g/mol) 309.83
Kvantitet 25 mg
Formel vikt 309.1284 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.