Läs mer
Demethyldihydrolanosterol, TRC
Produktkod. 30363608
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363608 10 mg
30356915 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363608 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23090510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5241-24-7
Molekylformel C29H50O
Molekylvikt (g/mol) 414.71
Synonym (3beta,5alpha)-4,4-Dimethylcholest-8-en-3-ol, 4,4-Dimethyl-5alpha-cholest-8-en-3beta-ol, 4,4-Dimethyl-5alpha-cholest-8(9)-en-3beta-ol
IUPAC-namn (3S,5R,10S,13R,14R,17R)-4,4,10,13-tetramethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,3,5,6,7,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2C3=C(CC[C@]12C)[C@@]4(C)CC[C@H](O)C(C)(C)[C@@H]4CC3
Specifikationer
Kemiskt namn eller material Demethyldihydrolanosterol
Kvantitet 10 mg
InChI formel InChI=1S/C29H50O/c1-19(2)9-8-10-20(3)22-12-13-23-21-11-14-25-27(4,5)26(30)16-18-29(25,7)24(21)15-17-28(22,23)6/h19-20,22-23,25-26,30H,8-18H2,1-7H3/t20-,22-,23+,25+,26+,28-,29-/m1/s1
Formel vikt 414.3862