Läs mer
Demethylpiperazinyl Desethyl Sildenafil Sulfonyl Chloride, TRC
Produktkod. 30355544
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30355544 25 mg
1 options
Produktkod. 30355544 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23130025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Demethylpiperazinyl Desethyl Sildenafil Sulfonyl Chloride
CAS 139756-27-7
Molekylformel C15H15ClN4O4S
Kvantitet 25 mg
Synonym 3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-hydroxy-benzenesulfonyl Chloride, 5-(5-Chlorosulfonyl-2-hydroxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
InChI formel InChI=1S/C15H15ClN4O4S/c1-3-4-10-12-13(20(2)19-10)15(22)18-14(17-12)9-7-8(25(16,23)24)5-6-11(9)21/h5-7,17H,3-4H2,1-2H3,(H,18,22)/b14-9+
LEDER CCCC1=NN(C2=C1N/C(=C\3/C=C(C=CC3=O)S(=O)(=O)Cl)/NC2=O)C
IUPAC-namn (3E)-3-(1-methyl-7-oxo-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-5-ylidene)-4-oxocyclohexa-1,5-diene-1-sulfonyl chloride
Molekylvikt (g/mol) 382.82
Formel vikt 382.0503