Läs mer
Denatonium Chloride, TRC
Produktkod. 30357819
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30357819 100 mg
1 options
Produktkod. 30357819 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD231708100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Denatonium Chloride
CAS 1674-99-3
Molekylformel C21H29ClN2O
Kvantitet 100 mg
Synonym Lignocaine Benzyl Chloride, Benzyldiethyl[(2,6-xylylcarbamoyl)methyl]ammonium Chloride, N-[2-[(2,6-dimethylphenyl)amino]-2-oxoethyl]-N,N-diethyl-benzenemethanaminium Chloride, Benzyldiethyl[(2,6-xylylcarbamoyl)methyl]benzenemethanaminium Chloride
InChI formel InChI=1S/C21H28N2O.ClH/c1-5-23(6-2,15-19-13-8-7-9-14-19)16-20(24)22-21-17(3)11-10-12-18(21)4;/h7-14H,5-6,15-16H2,1-4H3;1H
LEDER [Cl-].CC[N+](CC)(CC(=O)Nc1c(C)cccc1C)Cc2ccccc2
Molekylvikt (g/mol) 360.92
Formel vikt 360.1968