Läs mer
Deoxycholic Acid 3-O-Beta-D-Glucuronide Disodium Salt, TRC
Produktkod. 30007665
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007665 1 mg
30011366 10 mg
30005590 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2327201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59274-67-8
Molekylformel C30H46Na2O10
Molekylvikt (g/mol) 612.66
Synonym (3α,5β,12α)-23-Carboxy-12-hydroxy-24-norcholan-3-yl-β-D-glucopyranosiduronic Acid Disodium Salt;
LEDER C[C@@]12[C@](CC[C@]2([H])[C@@H](CCC(O[Na])=O)C)([H])C3CC[C@]4([H])C[C@H](O[C@@H]5O[C@H](C(O[Na])=O)[C@@H](O)[C@H](O)[C@H]5O)CC[C@]4(C)C3C[C@@H]1O
Specifikationer
Kemiskt namn eller material Deoxycholic Acid 3-O-beta-D-Glucuronide Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C30H48O10.2Na/c1-14(4-9-22(32)33)18-7-8-19-17-6-5-15-12-16(10-11-29(15,2)20(17)13-21(31)30(18,19)3)39-28-25(36)23(34)24(35)26(40-28)27(37)38;;/h14-21,23-26,28,31,34-36H,4-13H2,1-3H3,(H,32,33)(H,37,38);;/q;2*+1/p-2/t14-,15-,16-,17?,18-,19+,20?,21+,23+,24+,25-,26+,28-,29+,30-;;/m1../s1
Procent renhet >95%