Läs mer
Deoxycorticosterone 21-Glucoside, TRC
Produktkod. 30361901
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361901 50 mg
30352210 5 mg
30356634 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30361901 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23270850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4319-56-6
Molekylformel C27H40O8
Molekylvikt (g/mol) 492.6
Synonym Deoxycorticosterone Glucoside, Deoxycorticosterone beta-D-Glucopyranoside, 21-(beta-D-Glucopyranosyloxy)pregn-4-ene-3,20-dione, 11-Deoxycorticosterone 21-Glucoside, 11-Deoxycorticosterone Glucoside
LEDER C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O
Specifikationer
Kemiskt namn eller material Deoxycorticosterone 21-Glucoside
Kvantitet 50 mg
InChI formel InChI=1S/C27H40O8/c1-26-9-7-15(29)11-14(26)3-4-16-17-5-6-19(27(17,2)10-8-18(16)26)20(30)13-34-25-24(33)23(32)22(31)21(12-28)35-25/h11,16-19,21-25,28,31-33H,3-10,12-13H2,1-2H3
Formel vikt 492.2723