Läs mer
Deoxyfuconojirimycin, Hydrochloride, TRC
Produktkod. 30366809
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366809 100 mg
30355222 5 mg
30366600 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366809 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD236000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 210174-73-5
Molekylformel C6 H13 N O3 . Cl H
Molekylvikt (g/mol) 183.63
Synonym 3,4,5-Piperidinetriol, 2-methyl-, hydrochloride (1:1), (2S,3R,4S,5R)-, 3,4,5-Piperidinetriol, 2-methyl-, hydrochloride, (2S,3R,4S,5R)- (9CI), 1,5-Dideoxy-1,5-imino-L-fucitol hydrochloride, Deoxyfuconojirimycin hydrochloride, (2S,3R,4S,5R)-2-Methyl-3,4,5-trihydroxypiperidine hydrochloride, (2S,3R,4S,5R)-2-Methylpiperidine-3,4,5-triol hydrochloride
IUPAC-namn (2S,3R,4S,5R)-2-methylpiperidine-3,4,5-triol;hydrochloride
LEDER Cl.C[C@@H]1NC[C@@H](O)[C@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Deoxyfuconojirimycin, Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C6H13NO3.ClH/c1-3-5(9)6(10)4(8)2-7-3;/h3-10H,2H2,1H3;1H/t3-,4+,5+,6-;/m0./s1
Formel vikt 183.0662
Procent renhet >95%